Learn More
Texwipe™ TexVantage™ Polyester Cleanroom Wipers
TexVantage™ polyester cleanroom wipers are contstructed from 100% polyester, creating a wiper detail ideal for spill control, cleaning and solution application.
Manufacturer:  Texwipe™ TX8944
 View more versions of this product
Catalog No. 18-366-226
Add to cart
Description
Description
Polyester cleanroom wipes are designed for use on abraisive surfaces. This wiper will not easily snag or abrade releasing particles and fibers into the process or environment
Applications
  • Wiping and cleaning surfaces, equipment and parts
  • Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
  • Lining trays for holding, protecting, drying and storing of parts, equipment and devices
  • Appropriate for use with temperatures less than 400°F (250°C)
Polyester cleanroom wipes are designed for use on abraisive surfaces.  This wiper will not easily snag or abrade releasing particles and fibers into the process or environment
Applications
  • Wiping and cleaning surfaces, equipment and parts
  • Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
  • Lining trays for holding, protecting, drying and storing of parts, equipment and devices
  • Appropriate for use with temperatures less than 400°F (250°C)
Specifications
Specifications
4 x 4in.
100% Continuous-filament Polyester
6000 wipers total; 300 wipers/bag; 20 bags/case
Spill control, cleaning and solution application
Cleanroom wipers