Learn More
Texwipe™ TexVantage™ Polyester Cleanroom Wipers
TexVantage™ polyester cleanroom wipers are contstructed from 100% polyester, creating a wiper detail ideal for spill control, cleaning and solution application.
$24.50 - $31.80
Specifications
For Use With (Application) Spill control, cleaning and solution application
Material 100% Continuous-filament Polyester
Product Type Cleanroom wipers
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Packaging Quantity Price Quantity & Availability  
18-366-226
View Documents
Texwipe™
TX8944
4 x 4in. 6000 wipers total; 300 wipers/bag; 20 bags/case Pack of 300 for $31.80
Case of 20 PK for $540.00
Add to cart
18-366-225
View Documents
Texwipe™
TX8949
9 x 9in. 1500 wipers total; 150 wipers/bag; 10 bags/case Pack of 150 for $24.50
Case of 10 PK for $239.21
Add to cart
18-366-224
View Documents
Texwipe™
TX8942
12 x 12in. 1000 wipers total; 100 wipers/bag; 10 bags/case Pack of 100 for $30.30
Case of 10 PK for $297.00
Add to cart
Description
Description
Polyester cleanroom wipes are designed for use on abraisive surfaces. This wiper will not easily snag or abrade releasing particles and fibers into the process or environment
Applications
  • Wiping and cleaning surfaces, equipment and parts
  • Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
  • Lining trays for holding, protecting, drying and storing of parts, equipment and devices
  • Appropriate for use with temperatures less than 400°F (250°C)
Polyester cleanroom wipes are designed for use on abraisive surfaces.  This wiper will not easily snag or abrade releasing particles and fibers into the process or environment
Applications
  • Wiping and cleaning surfaces, equipment and parts
  • Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
  • Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone and degreasers
  • Lining trays for holding, protecting, drying and storing of parts, equipment and devices
  • Appropriate for use with temperatures less than 400°F (250°C)
Specifications
Specifications
Spill control, cleaning and solution application
100% Continuous-filament Polyester
Cleanroom wipers