Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ Alpha™ 10 Sealed-Border Wipers Get IT Item Available on GSA/VA Contract for Federal Government customers only.

For ISO Class 3 to 4 applications

Manufacturer:  Texwipe™ TX 1010

 View more versions of this product

Catalog No. 18-323A



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

320
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
9 x 9 in. (22.9 x 22.9cm)
9 in.
Dry Wiper
9 in.
Ideal wipes for use in multiple environments
22.9cm
130
Double-knit
Sealed
Polyester
9 in.
22.9cm
9 in.
Alpha™ 10 dry wipes
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit