Texwipe™ AlphaMop™ Isolator Cleaning Tool
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
Manufacturer: Texwipe™ TX7104
 View more versions of this product
Catalog No. 19-139-692
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For cleanroom class 4 to 8
    Head Type Swivel
    Includes Two handles one polyester foam pad and six TechniCloth™ polyester mop covers, mop with telescoping handle includes six polyester mop covers
    Autoclavable Yes
    Material Polyester
    Product Type Cleanroom Mop
  • Description & Specifications
Related Products
Product Content Feedback