Texwipe™ BetaMop™ Polyester Strip Refill Heads For Cleanroom Mops Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Texwipe™ STX7072
 View more versions of this product
Catalog No. 19-222-378
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Cleanroom Mops
    Autoclavable Yes
    Material Polyester
    Product Type Polyester Strip Refill Head
    Sterile Yes
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
  • Description & Specifications
Related Products
Product Content Feedback