Learn More
Texwipe™ BruClean™ Tablet Disinfectant Available on GSA/VA Contract for Federal Government customers only.
Convenient tablet form — easy to use
Manufacturer:  Texwipe™ TX6466
Catalog No. 18-368-140
Add to cart
Description
Description
BruClean TbC™ is a premeasured tablet that, when added to one gallon of water, creates a disinfectant cleaner that is effective against a broad array of pathogens including MRSA, E. coli and Salmonella.
  • Active ingredient is sodium dichloroisocyanurate (NaDCC)
  • Tablet is more stable than bleach, almost neutral in pH and biodegradable
  • Consistent strength produced at point of use for cleaning and disinfecting hard surfaces
  • Neutral pH makes it less corrosive on surfaces than liquid bleach
  • Pre-scored for half-strength
Cleaning and disinfecting hard surfaces, such as stainless steel; disinfecting all work areas — equipment, hoods, carts, isolators and floors; can easily replace liquid bleach in all applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
This product is EPA-registered (71847-2-106)
Specifications
Specifications
5 to 6
Sodium Dichloroisocyanurate
Cleaning and disinfecting hard surfaces such as Stainless Steel, work areas equipment, hoods, carts, isolators, floors etc.
EPA
Bottle
Tablet
Safety and Handling
Safety and Handling
ShelfLife : 2 Year(s)