Learn More
Texwipe™ Gloss Cleanroom Labels Available on GSA/VA Contract for Federal Government customers only.
Designed for critical environments where ink adhesion and reflectivity are desired
Manufacturer:  Texwipe™ TX542G
 View more versions of this product
Catalog No. 19-150-5602
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Permanent
Designed for critical environments where ink adhesion and reflectivity are desired, tested for surface particles, cleaned and double bagged
4 x 2 in.
-20 to 176°F
2.6mil
Polypropylene
Gloss
2 Years
10.16 x 5cm
-29 to 80°C
Label
1000 Labels/Roll