Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Supplier: Texwipe™ AMA364681W
Description
Apply directly to floor
- Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
- Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
White | |
30 Sheets/Mat | |
46 x 36 in. |