Learn More
Texwipe™ KOGO™ TPA Cleanroom Adhesive Tapes Available on GSA/VA Contract for Federal Government customers only.
Cleanroom tape solution for sealing and packaging material in critical environments
Manufacturer:  Texwipe™ TPA0596GR
 View more versions of this product
Catalog No. 18-366-404
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Acrylic
ISO 3
36 yd. x 0.5 in. (33m x 1.3cm)
Yes
33m
-40 to 88°C
0.5 in.
3 in.
96 Roll(s)
Green
7.62cm
Cleanroom packaging, color coding, labeling and waferbox sealing
36 yd.
-40 to 190°F
5.5mil
1.3cm
LDPE (Low-density Polyethylene)