Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Texwipe™ KOGO™ TPR Cleanroom Adhesive Tapes Available on GSA/VA Contract for Federal Government customers only.
For quick tack and moderate adhesion
Manufacturer:  Texwipe™ TPR3016GR
 View more versions of this product
Catalog No. 18-366-444
Add to cart
Specifications Description & Specifications
Specifications
Rubber,Synthetic
ISO 3
Ideal for cleanroom packaging, color coding, labeling and waferbox sealing
36 yd.
-40 to 190°F
5.5mil
3 in.
16 Roll(s)
Green
7.6cm
Yes
33m
-40 to 88°C
3 in.
LDPE (Low-density Polyethylene)