Learn More
Texwipe™ KOGO™ TVR Cleanroom Adhesive Tapes
Ideal for clean build or cleanroom construction applications where quick tack and abrasion resistance are important
Manufacturer:  Texwipe™ TVR2024YE
 View more versions of this product
Catalog No. 18-366-451
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Bonds quickly to irregular surfaces such as duct work or visqueen sheets used for temporary walls
  • Matte finish is easy to write on with a pen for labeling
  • Good for color coding or aisle marking in cleanrooms
  • PVC backing material with synthetic rubber adhesive
  • Roll length: 33m (36 yd.)
  • Various colors and widths available
Order Info
Packaged in press-and-seal cleanroom bags
Specifications
Specifications
Rubber, Synthetic
ISO 3
36 yd. x 2 in. (33m x 5.0cm)
Ideal for aisle marking, color coding, HEPA filter protection and cleanroom build
36 yd.
-20 to 176°F
6 mil
5.0 cm
PVC
Yellow
7.6 cm
Matte
Yes
33 m
-29 to 80°C
2 in.
3 in.
24 Roll(s)