Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ Polysat™ Presaturated Polypropylene Wipers Available on GSA/VA Contract for Federal Government customers only.

For ISO Class 6 to 7 applications

Manufacturer:  Texwipe™ TX1051

 View more versions of this product

Catalog No. 19-033-016



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

99
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
9 in.
50/Pk.
No
9 in.
Designed for cleaning equipment and other environmental surfaces during and following the production flow, suitable for wiping the exterior of production equipment in controlled environments
27.9cm
35
11 x 9 in. (27.9 x 22.8cm)
Polypropylene
70% Isopropyl Alcohol, 30% Deionised Water
11 in.
22.8cm
11 in.
PolySat™ prewetted wipes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit