Learn More
Texwipe™ SolvSat™ Canister System
A controlled delivery system for prewetted wipers to deliver the right amount of solvent to manual cleaning operations
Manufacturer:  Texwipe™ TX8344
Catalog No. 18-366-463
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Designed for ease of use in a production environment, system can be easily carried to the workstation. Each pouch contains a perforated roll of nonwoven Rayon wipers saturated with a 99% toluene solution.Wipers are packaged in a solvent-resistant pouch that fits easily into a canister. Wipes are threaded through a molded opening designed to allow users to tear off wipes at their perforation for single wipe use.
Features:
  • Durable, chemical resistant wipers
  • Wipers prewet with accurately measured volumes of toluene
  • Color coded label for easy product identification
  • Lot coded for traceability
Benefits:
  • Eliminates step of adding solvent to rags
  • Minimizes worker exposure to hazardous chemicals
  • Optimizes solvent usage
  • Reduces spill potential and VOC emissions
  • Eliminates potential for cross contamination of squirt bottles
Compliance
QPL Listed AMS3819; Meets Aerospace Standard BMS 15-5F, Class A; Meets Aerospace Standard DMS 1820E
Specifications
Specifications
AMS 3819, BMS 15-5F, DMS 1820E
Cleaning of printing plates, blankets, rollers and tools
23 cm
750mL 99% Toluene
15 cm
9 x 6 in. (23 x 15 cm)
9 in.
SolvSat™ canister system
6 in.