Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ TechniCloth™ II Blended Wipers Available on GSA/VA Contract for Federal Government customers only.

For ISO Class 6 and 7 applications

Manufacturer:  Texwipe™ TX 1109

 View more versions of this product

Catalog No. 18-330A



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

260
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
9 in.
9 in.
22.9cm
9 in.
TechniCloth™ II dry wipes
67
9 x 9 in. (22.9 x 22.9cm)
Polyester, Cellulose
9 in.
Designed for critical environments where low particle generation and high absorbency are prime considerations, excellent for use in facilities for integrated circuit, printed circuit board and electronic components fabrications
22.9cm
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit