Learn More
Texwipe™ Vectra™ Alpha™ Nu Knit Wipers
For use in ISO Class 1 to 4 cleanrooms
Manufacturer:  Texwipe™ TX1069
Catalog No. 19-168-473
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Engineered for high-sorbency use in critical cleanroom environments, such as semiconductor and pharmaceutical manufacturing.
  • No-run interlock, 100% continuous-filament virgin polyester construction lowers contamination risk
  • No barrier film left behind compared to standard polyester wipers
  • Border sealed with smooth surface, no rough areas on edge of fibers that can snag and easily rupture releasing particles
  • Very low particle counts and NVRs
Specifications
Specifications
400
9 x 9 in. (23 x 23 cm)
9 in.
Polyester
Super sealed
9 in.
23 cm
100% Continuous filament
For ISO Class 1 to 4 cleanroom most critical cleanroom environment applications
23 cm
Vectra™ Alpha™ Nu knit wipes
9 in.
9 in.