Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAM Monoclonal Antibody (HL2012), Invitrogen™

Rabbit Monoclonal Antibody

Supplier:  Thermo Scientific MA547258

Encompass

Catalog No. PIMA547258


Add to Cart

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

TFAM Monoclonal antibody specifically detects TFAM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFAM
Monoclonal
1 mg/mL
PBS with no preservative
Q00059
Tfam
Full length human mtTFA recombinant protein
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL2012
Unconjugated
Tfam
AI661103; Hmgts; Mitochondrial transcription factor 1; mitochondrial transcription factor A; MTTF1; mtTFA; TCF6; TCF-6; TCF6L1; TCF6L2; TCF6L3; Testis-specific high mobility group protein; testis-specific HMG-box protein m-tsHMG; TFAM; Transcription factor 6; transcription factor 6-like 1; transcription factor 6-like 2; transcription factor 6-like 2 (mitochondrial transcription factor); transcription factor 6-like 3; transcription factor A, mitochondrial; tsHMG; TS-HMG
Rabbit
Protein A
RUO
7019
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit