Learn More
TFAP2D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188351AP
Catalog No. 50-173-2251
Add to cart
Description
Description
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5 ft.-GCCNNNGGC-3 ft. and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.
Specifications
Specifications
TFAP2D
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q7Z6R9, Q91ZK0
AP2 delta, TFAP2BL1, TFAP2D
Rabbit
IgG
150 μL
-20°C
Primary
226896, 301284, 83741
Western Blot
Unconjugated
TFAP2D
Liquid
Tfap2d
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat