Learn More
TFB2M Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 244111AP
Catalog No. 50-173-4340
Add to cart
Description
Description
S-adenosyl-L-methionine-dependent methyltransferase which specifically dimethylates mitochondrial 12S rRNA at the conserved stem loop. Also required for basal transcription of mitochondrial DNA, probably via its interaction with POLRMT and TFAM. Stimulates transcription independently of the methyltransferase activity. Compared to TFB1M, it activates transcription of mitochondrial DNA more efficiently, while it has less methyltransferase activity.
Specifications
Specifications
TFB2M
0.62 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TL26, Q9H5Q4
h mtTFB, h mtTFB2, Hkp1, hTFB2M, mtTFB2, NS5ATP5, TFB2M
Rabbit
IgG
150 μL
-20°C
Primary
15278, 64216
Western Blot
Unconjugated
TFB2M
Liquid
TFB2M
TFB2M Fusion Protein Ag19687
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse