Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1 Mouse anti-Human, Clone: SPM313, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human TFF1.

Manufacturer:  Abnova Corporation MAB13166

Catalog No. 89-934-996


Add to cart

Description

Description

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq]

Sequence: CFDDTVRGVPWCFYPNTIDVPPEEECEF
Specifications

Specifications

TFF1
SPM313
Mouse monoclonal antibody raised against synthetic peptide of human TFF1.
In 10mM PBS.
BCEI/D21S21/HP1.A/HPS2/pNR-2/pS2
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TFF1
A synthetic peptide corresponding to amino acids 57-84 at C-terminus region of human TFF1.
Protein A/G purification
RUO
Primary
7031
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit