Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1/pS2 Mouse anti-Human, Clone: rTFF1/1091, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen TFF1/pS2
Clone rTFF1/1091
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122158
View Documents
Novus Biologicals
NBP30734220UG
20 μg
Each for $159.82
Add to cart
 
NB122157
View Documents
Novus Biologicals
NBP30734210UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TFF1/pS2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A
Recombinant full-length human TFF1/pS2 protein
Monoclonal
Protein A or G purified
rTFF1/1091
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Breast Cancer, Cancer
10 mM PBS with 0.05% BSA
7031
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit