Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF2 Mouse anti-Human, Clone: GE16C, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen TFF2
Clone GE16C
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122174
View Documents
Novus Biologicals
NBP30735020UG
20 μg
Each for $138.37
Add to cart
 
NB122173
View Documents
Novus Biologicals
NBP30735010UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

TFF2 Monoclonal antibody specifically detects TFF2 in Human samples. It is validated for ELISA
Specifications

Specifications

TFF2
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
7032
IgM Kappa
Primary
Store at 4°C.
GE16C
ELISA
Unconjugated
Mouse
Human
SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, spasmolysin, Spasmolytic polypeptide, trefoil factor 2
Synthetic peptide corresponding to a 16-amino acid C-terminal sequence of the human TFF2.
Monoclonal
Protein L purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit