Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFG Mouse anti-Human, Clone: TFG-03, Abnova™
Mouse monoclonal antibody raised against recombinant human TFG.
Supplier: Abnova Corporation MAB12858
Specifications
TFG | |
Monoclonal | |
Unconjugated | |
Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user. | |
FLJ36137/TF6/TRKT3 | |
Mouse | |
Protein A purification | |
RUO | |
10342 | |
Store at 4°C. Do not freeze. | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
TFG-03 | |
Mouse monoclonal antibody raised against recombinant human TFG. | |
In PBS, pH 7.4 (0.09% sodium azide). | |
TFG | |
Recombinant Trx-His-tag fusion protein corresponding to N-terminus region of human TFG. | |
50 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction