Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRK fused gene Monoclonal Antibody (OTI2C3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525759
Description
TRK fused gene Monoclonal antibody specifically detects TRK fused gene in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TRK fused gene | |
Monoclonal | |
0.45 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92734 | |
TFG | |
Full length human recombinant protein of TFG produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C3 | |
Unconjugated | |
TFG | |
AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene | |
Mouse | |
Affinity Chromatography | |
RUO | |
10342 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction