Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFIISH Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19131820UL

 View more versions of this product

Catalog No. NBP19131820


Add to cart

Description

Description

TFIISH Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

TFIISH
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TCEA3
The specific Immunogen is proprietary information. Peptide sequence LKDPRNPGLRRNVLSGAISPELIAKMTAEEMASDELRELRNAMTQEAIRE.
Immunogen affinity purified
RUO
Primary
6920
Western Blot
Western Blot 1:1000
NP_035672
rhabdomyosarcoma antigen MU-RMS-40.22, TFIIS, TFIIS.H, TFIISH, transcription elongation factor A (SII), 3, transcription elongation factor A protein 3, Transcription elongation factor S-II protein 3, Transcription elongation factor TFIIS.h
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit