Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFIP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549275
Description
Predicted to react with monkey based on sequence homology.
Involved in pre-mRNA splicing, specifically in spliceosome disassembly during late-stage splicing events. Intron turnover seems to proceed through reactions in two lariat-intron associated complexes termed Intron Large (IL) and Intron Small (IS). In cooperation with DHX15 seems to mediate the transition of the U2, U5 and U6 snRNP-containing IL complex to the snRNP-free IS complex leading to efficient debranching and turnover of excised introns. May play a role in the differentiation of ameloblasts and odontoblasts or in the forming of the enamel extracellular matrix.
Specifications
TFIP11 | |
Polyclonal | |
Unconjugated | |
TFIP11 | |
2810002G02Rik; AF097181; AW046167; bK445C9.6; hNtr1; HSPC006; NTR1; Septin and tuftelin-interacting protein 1; Spp382; Srr1; STIP; STIP-1; Tfip11; TIP33; TIP39; tuftelin interacting protein 11; tuftelin-interacting protein 11; tuftelin-interacting protein 33; tuftelin-interacting protein 39; tuftelin-interacting protein, 39 kD | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
24144 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9UBB9 | |
TFIP11 | |
KLH conjugated synthetic peptide between 42-70 amino acids from the N-terminal region of human TFIP11. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction