Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ TFM™ Colored Tissue Freezing Medium
SDP

Variety of colors to easily differentiate multiple specimens during sectioning and staining

$89.00 - $119.00

Specifications

Boiling Point 212°C
Product Type Tissue Freezing Medium
Quantity 118mL
For Use With (Application) Embedding matrix for frozen sections
View More Specs

Products 6
Catalog Number Mfr. No. Color Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Color Product Type Price Quantity & Availability  
15-183-13
SDP
View Documents
General Data Healthcare™
TFMC
Clear Tissue Freezing Medium
Each for $89.00
Case of 12 EA for $888.00
Add to Cart
 
15-183-41
SDP
View Documents
General Data Healthcare™
TFMY
Yellow Tissue Freezing Medium
Each for $119.00
Case of 4 EA for $394.00
Add to Cart
 
15-183-37
SDP
View Documents
General Data Healthcare™
TFMB
Blue Tissue Freezing Medium
Each for $119.00
Case of 4 EA for $394.00
Add to Cart
 
15-183-40
SDP
View Documents
General Data Healthcare™
TFMR
Red Tissue Freezing Medium
Each for $119.00
Case of 4 EA for $394.00
Add to Cart
 
15-183-39
SDP
View Documents
General Data Healthcare™
TFMG
Green Tissue Freezing Medium Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
15-183-36
SDP
View Documents
General Data Healthcare™
TFM5
Blue, Clear, Green, Red, and Yellow Tissue Freezing Medium Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »
Description

Description

  • Unique formula reduces curling, allowing easier pickup of flat serial sections
  • Reduced water content minimizes freeze fractures

Order Info

Supplied in 4 oz. (118mL) plastic squeeze bottles. Complete set includes one of all five medium colors in a wooden rack.

Specifications

Specifications

212°C
Tissue Freezing Medium
118mL
Embedding matrix for frozen sections
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit