Learn More
TFPI-2 Mouse anti-Human, Alexa Fluor 488, Clone: MM0564-9F6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211918AF488
Catalog No. NB211918488
Add to cart
Description
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
TFPI-2
MM0564-9F6
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
TFPI2
Human TFPI2
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Placental protein 5, PP5FLJ21164, TFPI-2REF1, tissue factor pathway inhibitor 2
Mouse
IgG1
0.1 ml
Primary
7980