Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ TFPI Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Supplier: Novus Biologicals™ NBP27953115PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TFPI.Specifications
24 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Mouse | |
convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
TFPI | |
RUO | |
TFPI |
23.44-1500 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TFPI. | |
Sandwich ELISA | |
7035 | |
Rabbit | |
15 Plates | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1.5 ng/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction