Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFPI Mouse anti-Human, Clone: OTI4H9, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF806052
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predomitly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.Specifications
TFPI | |
Monoclonal | |
Unconjugated | |
TFPI | |
Lyophilized | |
TFPI | |
Human recombit protein fragment corresponding to amino acids 29-304 of human TFPI produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P10646 | |
EPI, LACI, TFI, TFPI1 | |
Mouse | |
IgG2a | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
7035 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title