Learn More
TFPI Mouse anti-Human, Clone: OTI1E1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805675
Catalog No. 50-166-7532
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predomitly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.
Specifications
Specifications
TFPI
Monoclonal
Unconjugated
TFPI
Lyophilized
TFPI
Human recombit protein fragment corresponding to amino acids 29-304 of human TFPI produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1E1
PBS with 8% trehalose and no preservative; pH 7.3
P10646
EPI, LACI, TFI, TFPI1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7035