Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFPI Mouse anti-Human, Clone: OTI4H9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526775

Catalog No. PIMA526775


Add to cart

Description

Description

Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predomitly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.
Specifications

Specifications

TFPI
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P10646
tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor); TFPI; EPI; LACI; TFI; TFPI1; OTTHUMP00000163472; OTTHUMP00000205431; anti-convertin; extrinsic pathway inhibitor; lipoprotein-associated coagulation inhibitor; tissue factor pathway inhibitor
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7035
Immunohistochemistry (Paraffin), Western Blot
OTI4H9
Unconjugated
TFPI
Liquid
TFPI
Human recombit protein fragment corresponding to amino acids 29-304 of TFPI produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit