Learn More
TFPI2 Mouse anti-Human, Clone: OTI5H3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805493
Catalog No. 50-166-7452
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
Specifications
Specifications
TFPI2
Monoclonal
Unconjugated
TFPI2
Lyophilized
TFPI2
Human recombit protein fragment corresponding to amino acids 23-235 of human TFPI2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5H3
PBS with 8% trehalose and no preservative; pH 7.3
P48307
PP5, REF1, TFPI-2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7980