Learn More
TfR (Transferrin R) Mouse anti-Human, Mouse, Rat, Primate, DyLight 680, Clone: 1A1B2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237317FR
Catalog No. NBP237317FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TfR (Transferrin R)
1A1B2
Western Blot, ELISA
Purified
TFRC
Purified recombinant fragment of human TFRC (AA: 608-727) expressed in E. Coli.
Protein A or G purified
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary
7037
ELISA, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Primate, Rat