Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Mouse anti-Human, PE-Cyanine7, Clone: TFRC/1818, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254466PECY7

Catalog No. N25446PECY7


Add to cart

Description

Description

TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TfR (Transferrin R)
TFRC/1818
Flow Cytometry
Conjugated, Purified
TFRC
Recombinant human extracellular TFRC protein fragment (aa94-212) (exact sequence is proprietary)
Protein A or G purified
Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary
7037.0
Human
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
It recognizes a ∽90-95kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit