Learn More
TfR (Transferrin R) Mouse anti-Rat, Alexa Fluor™ 750, Clone: OX26, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10079S100UG
Catalog No. FB10079S100
Add to cart
This item is not returnable. View return policy
Description
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
TfR (Transferrin R)
OX26
Flow Cytometry 0.25-1 μg/106 cells
Purified, Supernatant, Conjugated
Mouse
IgG2a
100 μg
Primary
7037
Rat
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
PHA activated rat lymphocytes
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects rat TfR in Flow Cytometry.