Learn More
TFR2 Mouse anti-Human, Mouse, Rat, Clone: OTI2B4, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504563
Catalog No. 50-167-5221
Add to cart
Description
Description
This gene is a member of the transferrin receptor-like family and encodes a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. This protein mediates cellular uptake of transferrin-bound iron and mutations in this gene have been associated with hereditary hemochromatosis type III. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
Specifications
TFR2
Monoclonal
Unconjugated
TFR2
Liquid
Tfr2
Full length human recombit protein of human TFR2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
B2GUY2, Q9JKX3, Q9UP52
HFE3, TFRC2
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
288562, 50765, 7036