Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFRC Mouse anti-Human, Clone: TFRC/1059, Abnova™

Mouse monoclonal antibody raised against full length recombinant human TFRC.

Manufacturer:  Abnova Corporation MAB13064

Catalog No. 89-934-998


Add to cart

Description

Description

Specifications

Specifications

TFRC
TFRC/1059
Mouse monoclonal antibody raised against full length recombinant human TFRC.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CD71/TFR/TFR1/TRFR
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
This antibody is highly specific to TFRC and shows no cross-reaction with other related proteins.
Flow Cytometry, Immunofluorescence
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TFRC
Recombinant protein corresponding to full length of human TFRC.
Protein A/G purification
RUO
Primary
7037
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit