Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFRC Mouse anti-Human, Clone: TFRC/1817, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15003

Catalog No. 89-035-023


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human TFRC.

  • High specificity
  • High reactivity
Specifications

Specifications

TFRC
TFRC/1817
Mouse monoclonal antibody raised against partial recombinant human TFRC.
In 10 mM PBS.
Liquid
TFRC
Recombinant protein corresponding to amino acids 94-212 of human TFRC.
Protein A/G purification
RUO
Primary
7037
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (2-4 ug/mL for coating) Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optima
P02786
CD71|TFR|TFR1|TRFR
Mouse
IgG2b, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)