Learn More
TG 100713, Tocris™
PI 3-kinase inhibitor
Manufacturer:  R&D Systems 4264/10
Quantity 10mg
 View more versions of this product
Catalog No. 42-641-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
254.25
TG 100713
C12H10N6O
3-(2, 4-Diamino-6-pteridinyl)-phenol
Specifications
10mg
>98%
254.25
TG 100713
925705-73-3
C12H10N6O
3-(2, 4-Diamino-6-pteridinyl)-phenol
Safety and Handling
Recommended Storage : Desiccate at Room Temperature