Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ TG 4-155

High affinity and selective EP2 receptor antagonist

Manufacturer:  Tocris™ 5052/50

 View more versions of this product

Catalog No. 50-525-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

High affinity and selective EP2 receptor antagonist (KB values are 2.4 and 34.5nM for EP2 and DP1 receptors respectively). Exhibits >500-fold selectivity for EP2 over other prostanoid receptors. Inhibits butaprost-induced proinflammatory cytokine production, cell proliferation and invasion of PC-3 cells in vitro.
  • Chemical Name: (2E)-N-[2-(2-Methyl-1H-indol-1-yl)ethyl]-3-(3,4,5-trimethoxyphenyl)-2-propenamide
  • Formula: C23H26N2O4
  • CAS Number: 1164462-05-8
  • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

TG 4-155
>98
50mg
TG 4-155
394.46
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit