Learn More
TGF-alpha Mouse anti-Human, Mouse, Rat, Bovine, Alexa Fluor™ 532, Clone: MF9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234683AF532
Catalog No. NP234683A32
Description
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human,Mouse,Rat,Bovine samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen).
Specifications
Specifications
TGF-alpha
MF9
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat, Bovine
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
TGFA
A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGFalpha.
0.1 mL
Primary
7039