Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha Mouse anti-Human, Mouse, Rat, Bovine, Clone: SPM357, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen TGF-alpha
Clone SPM357
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122181
View Documents
Novus Biologicals
NBP30735410UG
100 μg
Each for $352.89
Add to cart
 
NB122182
View Documents
Novus Biologicals
NBP30735420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

TGF-alpha
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat, Bovine
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
A 17-amino acid synthetic peptide from the carboxyl-terminus of rat TGF-alpha
Monoclonal
Protein A or G purified
SPM357
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Unconjugated
Mouse
Angiogenesis, Cell Cycle and Replication, Signal Transduction
10 mM PBS with 0.05% BSA
7039
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit