Learn More
TGF-alpha Mouse anti-Human, Rabbit, Zebrafish, Alexa Fluor™ 532, Clone: TG86, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234722AF532
Catalog No. NP234722A32
Add to cart
Description
Description
TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human,Rabbit,Zebrafish samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
TGF-alpha
TG86
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
TGFA
A 10-amino acid synthetic peptide (aa 34-43) from human TGFalpha.
0.1 mL
Primary
7039
Human, Rabbit, Zebrafish