Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha Rabbit anti-Human, Clone: 12, Novus Biologicals™
Rabbit Monoclonal Antibody
$340.00 - $521.00
Specifications
Antigen | TGF-alpha |
---|---|
Clone | 12 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for ELISA.Specifications
TGF-alpha | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
Recombinant Human TGF-alpha Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
12 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
0.2 um filtered solution in PBS | |
7039 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title