Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-alpha, Rabbit anti-Human, Clone: tAb2, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810820.025MG
Description
TGF-alpha Monoclonal specifically detects TGF-alpha in Human samples. It is validated for Flow Cytometry, ELISA, Dot Blot.Specifications
TGF-alpha | |
Monoclonal | |
1 mg/mL | |
ELISA, Dot Blot | |
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
tAb2 binds the N-terminal sequence of TGF-alpha, VVSHFND (with residues 4 to 7 being the minimal binding epitope). | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
ELISA, Dot Blot | |
tAb2 | |
Unconjugated | |
PBS | |
TGFA | |
Biologically active hemocyanin-coupled human TGF-alpha. | |
0.025 mg | |
Angiogenesis, Cell Cycle and Replication, Signal Transduction | |
7039 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction