Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGF-alpha, Rabbit anti-Human, Clone: tAb2, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen TGF-alpha
Clone tAb2
Applications ELISA, Dot Blot
Host Species Rabbit
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015618
View Documents
Novus Biologicals
NBP2810820.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015619
View Documents
Novus Biologicals
NBP2810820.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human Samples. It is validated for ELISA, Dot Blot.
Specifications

Specifications

TGF-alpha
ELISA, Dot Blot
Human
Primary
tAb2
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit