Learn More
TGF-beta 1 Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: 7F6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222114AF594
Catalog No. NP222114A94
Add to cart
Description
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
TGF-beta 1
7F6
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
TGFB1
Purified recombinant fragment of human TGF beta 1 (amino acids 62-197) expressed in E. coli. [UniProtNo. P01137]
0.1 mL
Primary
7040