Learn More
LAP (TGF-beta 1) Rat anti-Mouse, Alexa Fluor 647, Clone: 860206, R&D Systems
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB7666R100UG
Catalog No. FAB7666R100
Add to cart
This item is not returnable. View return policy
Description
Description
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
LAP (TGF-beta 1)
Monoclonal
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
CED, DPD1, LAP, LAP (TGFbeta 1), TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, transforming growth factor beta 1
Chinese hamster ovary cell line CHO-derived recombinant mouse TGF-beta 1 Met1-Ser390 Accession # P04202
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7040
Mouse
Flow Cytometry
860206
Flow Cytometry 0.25-1 ug/10^6 cells
P04202
Rat
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse LAP (TGF-beta 1) in flow cytometry.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)