Learn More
TGF-beta 2 Mouse anti-Human, Alexa Fluor 488, Clone: MM0640-6M28, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP232727AF488
Catalog No. NB232727488
Add to cart
Description
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
TGF-beta 2
MM0640-6M28
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
TGFB2
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human TGF-beta2
Protein G purified
Angiogenesis, Apoptosis, Biologically Active Proteins, Cell Cycle and Replication
Primary
7042
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Human