Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGF-beta 2 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0640-6M28, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP232727AF532
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TGF-beta 2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
TGFB2 | |
Human recombinant TGF-beta 2 | |
0.1 mL | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0640-6M28 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
Mouse | |
Protein G purified | |
Angiogenesis, Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
7042.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title